Learn More
Abbott™ Determine™ HIV-1/2 Ag/Ab Combo Test
Shop All Abbott Rapid Diagnostics Products
Click to view available options
Quantity:
25 Tests
Description
- In vitro, visually read, qualitative immunoassay gives faster results than conventional tests that rely solely on presence of HIV-1/2 antibodies
- Can be used with human serum, plasma, capillary (fingerstick) whole blood or venipuncture (venous) whole blood
- May distinguish acute HIV-1 infection from established HIV-1 infection when specimen is positive for HIV-1 p24 antigen and negative for anti-HIV-1 and anti-HIV-2 antibodies
- Suitable for use in multi-test algorithms designed for statistical validation of rapid HIV test results
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Clia Complexity | Waived |
Detectable Analytes | HIV-1/2 Antibodies, free HIV-1 p24 Antigen |
Sample Type | Fingerstick Whole Blood, Plasma, Serum, Venous Whole Blood |
Type | Determine™ HIV-1/2 Ag/Ab Combo Testing Kit |
Quantity | 25 Tests |
Test Time | 20 min. |